Orders LinkContactUsLinkComponent
Crosslinkers
Crosslinkers
Supplier:   CONSORT
Crosslinkers
Supplier:  CONSORT
Certificates
Crosslinkers
3 Options Available Below
Product Details & Documents
Three models: 254, 312, and 365 nm, each with 5× 8-watt UV tubes.
  • Nine presets for time and energy exposures as well as manual control by the user
  • Timer up to 999,9 minutes
  • Exposure ranges: 0 to 99,99 and 0 to 9,999 J/cm²
  • Storage of the last UV settings in a non-volatile memory
  • Photo-feedback system automatically compensates for output variation and tube aging
  • UV-blocking viewing window
Internal dimensions (W×D×H): 260×330×150 mm
External dimensions (W×D×H): 350×360×310 mm
Product Family Options
Product Information
  • Model
    Wavelength
    Pk
    Availability
    Price
  • CONSVLBLX-M
    Crosslinker
    Crosslinker
    E2922
    312 nm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 210,00 €
    3 210,00 €
    1 * 1 ST
       3 210,00 €
  • CONSVLBLX-C
    Crosslinker
    Crosslinker
    E2921
    254 nm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 210,00 €
    3 210,00 €
    1 * 1 ST
       3 210,00 €
  • CONSVLBLX-L
    Crosslinker
    Crosslinker
    E2923
    365 nm
    1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 210,00 €
    3 210,00 €
    1 * 1 ST
       3 210,00 €