Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
DNA QuantLadders

DNA QuantLadders

QuantLadder, 100 - 1000 bp, 50 applications, 50
Catalog # LONZ50334
Supplier:   LONZA
DNA QuantLadders
Catalog # LONZ50334
Supplier:  LONZA
CAS Number:  
DNA QuantLadders

QuantLadder, 100 - 1000 bp, 50 applications, 50

CAS Number:  
Price
227,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

QuantLadders are prepared from five highly purified DNA fragments that provide visual quantitation of DNA within ±1% accuracy. The DNA QuantLadder provides each DNA fragment in perfect relative mass equivalence at 100 ng, 70 ng, 50 ng, 20 ng and 10 ng. The reverse QuantLadder simply reverses the mass equivalence order of the QuantLadder.

  • Storage temperature: 2 to 8 °C
  • Also available in SimplyLoad® format
Specifications

Specifications

  • Cdt:
    50
  • Désignation:
    QuantLadder, 100 - 1000 bp, 50 applications

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.