ContactUsLinkComponent
VWR® Q path® Safesolv, Solvent Substitute
VWR® Q path® Safesolv, Solvent Substitute
Catalog # 00699464.
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Q path® Safesolv, Solvent Substitute
Catalog # 00699464.
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Description:
    Safesolv Q path® solvent substitute
Specifications
About this item
Q Path® Safesolv is a non hazardous replacement for xylene or toluene. It is compatible with the Q Path® Safemount mounting medium reference 00647520. It is used in place of xylene, toluene, methylecyclohexane or other solvents in the course of the dehydration stages of histological specimens and during the stages of staining and lamella mounting.
  • Clear colourless liquid