Membrane filtration set
Supplier: Hach
Jump to:
About this item
This membrane filtration set can be used for filtration of turbid samples for the determination of dissolved ingredients.
Delivery information: The set contains 50 syringe filters and one disposable 10 ml syringe.
2 Options Available Below
Product Details & Documents
This membrane filtration set can be used for filtration of turbid samples for the determination of dissolved ingredients.
Delivery information: The set contains 50 syringe filters and one disposable 10 ml syringe.
Product Family Options
Product Information
- Pore sizeAvailabilityPrice
- HACHLCW916Membrane filtration set including 50 syringe filters and 1 disposable 10 ml syringeMembrane filtration set including 50 syringe filters and 1 disposable 10 ml syringe0,45 µmLog in for availability1 * 50 ST212,00 €
- HACHLCW904Membrane filtration set including 50 syringe filters and 1 disposable 10 ml syringeMembrane filtration set including 50 syringe filters and 1 disposable 10 ml syringe1,20 µmLog in for availability1 * 50 ST201,00 €