Order Entry
Belgium
ContactUsLinkComponent
siRNA transfection reagent, RiboJuice™
siRNA transfection reagent, RiboJuice™
Catalog # 71115-4
CAS Number:  
siRNA transfection reagent, RiboJuice™
Catalog # 71115-4
CAS Number:  

Specifications

  • Pk:
    1 mL
  • siRNA:
    Yes
  • Description:
    RiboJuice™ siRNA transfection reagent, 1 ml

Specifications

About this item

RiboJuice™ siRNA Transfection Reagent delivers siRNA into a wide range of mammalian cell lines for targeted gene suppression. It is compatible with GeneJuice® Transfection Reagent for cotransfection of siRNA and plasmid DNA for normalising transfection efficiency with a reporter gene.

  • Highly flammable
  • Easy-to-use siRNA transfection reagent
  • Optimised and formulated for minimal toxicity
  • Used for gene suppression in a variety of cell lines successfully

Storage temperature: +2 to +8 °C.