Order Entry
Belgium
Orders LinkContactUsLinkComponent
Wall clock, classic

Wall clock, classic

Wall clock, classic, Black, 300 mm
Catalog # MOIN1179AC
Supplier:   MOINEAU
Wall clock, classic
Catalog # MOIN1179AC
Supplier:  MOINEAU
Wall clock, classic
Wall clock, classic

Wall clock, classic, Black, 300 mm

Price
53,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

ABS/mineral glass.

  • Black casing
  • For interior use
  • Without battery

Weight: 700 g

Specifications
  • Ø:
    300 mm
  • Colour:
    Black
  • Description:
    Wall clock, classic
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.