Orders LinkContactUsLinkComponent
Test strips, drug testing for urine
Test strips, drug testing for urine
Test strips, drug testing for urine
Supplier:  SURESCREEN
Certificates
Test strips, drug testing for urine
Test strips, drug testing for urine
5 Options Available Below
Product Details & Documents
Single test strips are an efficient method for testing urine for specific drugs in urine. Simply dip the test strip into a urine sample and read at 3 minutes. Each individual strip is provided in a foil pouch, to ensure the high quality of the product is retained throughout the shelf-life.
  • More than 20 different drug tests available
  • Easy to perform test routine
  • Clear results in 3 minutes
  • Greater than 99% accuracy
  • For detecting recent drug use approx. 1 to 8 days
Recommended markets and use:
On-site screening
Safety critical situations
Random screening
For-cause screening
Monitoring rehabilitation
Drug Type Options:
ACL - Clonazepam
AMP - Amphetamines
BAR - Barbiturates
BUP - Buprenorphine
BZO - Benzodiazepines
COC - Cocaine
EDDP - Methadone Metabolite
ETG - Ethyl Glucuronide
FEN - Fentanyl
K2 - Synthetic Cannabis (K2/Spice)
KET - Ketamine
MAMP - Methamphetamines
MDMA - Ecstasy
MOP - Morphine
MTD - Methadone
OPI - Opiates
OXY - Oxycodone
PCP - Phencyclidine
PPX - Propoxyphene
TCA - Tricyclic Antidepressants
TRA - Tramadol
THC - Cannabis
Documents
Product Family Options
Product Information
  • Sensitivity
    Availability
    Price
  • 700-9544
    Amphetamine drug test strip for urine
    Amphetamine drug test strip for urine
    1000 ng/ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 61,80 €
    61,80 €
    1 * 50 ST
       61,80 €
  • 720-0657
    Benzodiazepine drug test strip
    Benzodiazepine drug test strip
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 61,80 €
    61,80 €
    1 * 50 ST
       61,80 €
  • 700-9564
    Cannabis drug test strip
    Cannabis drug test strip
    50 ng/ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 61,80 €
    61,80 €
    1 * 50 ST
       61,80 €
  • 720-0737
    Methadone drug test strip
    Methadone drug test strip
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1,30 €
    1,30 €
    1 * 50 ST
       1,30 €
  • 700-9634
    Opiate drug test strip
    Opiate drug test strip
    2000 ng/ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 1,30 €
    1,30 €
    1 * 50 ST
       1,30 €