Anti-Trem-1 Rat Monoclonal Antibody [clone: L5-B8.2A12.3A12]
Catalog # SERTMCA2578GA
Supplier: Serotec
Specifications
- Antibody type:Primary
- Antigen symbol:TREM-1
- Clonality:Monoclonal
- Clone:L5-B8.2A12.3A12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:liquid
- Storage buffer:PBS
- Storage temperature:+4 °C, –20 °C if preferred
- Concentration:1,0 mg/ml
- Shipping temperature:4 °C
- Immunogen:Mouse TREM-1 Fc fusion protein
- Purification:Purified by affinity chromatography on Protein G from tissue culture supernatant
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Trem-1
Clonality: Monoclonal
Clone: L5-B8.2A12.3A12
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse