Order Entry
Belgium
ContactUsLinkComponent
 
Anti-CD68 (Macrosialin) Rat Monoclonal Antibody [clone: FA-11]
  SERTMCA1957
 :  Serotec
Anti-CD68 (Macrosialin) Rat Monoclonal Antibody [clone: FA-11]
  SERTMCA1957
 :  Serotec

 

  • Antibody type:
    Primary
  • Antigen name:
    Macrosialin
  • Antigen symbol:
    CD68
  • Clonality:
    Monoclonal
  • Clone:
    FA-11
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    liquid
  • Storage buffer:
    PBS
  • Storage temperature:
    +4 °C, –20 °C if preferred
  • Concentration:
    1,0 mg/ml
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by affinity chromatography on Protein G from tissue culture supernatant
  • Size:
    250 μg
  • Pk:
    0,25 mg

 

 

Type: Primary
Antigen: CD68 (Macrosialin)
Clonality: Monoclonal
Clone: FA-11
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse