Anti-CD11B (Integrin Alpha M Chain, Mac-1) Rat Monoclonal Antibody [clone: 5C6]
Catalog # SERTMCA711G
Supplier: Serotec
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha M
- Antigen symbol:ITGAM
- Clonality:Monoclonal
- Clone:5C6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human,Mouse
- Cross adsorption:No
- Format:liquid
- Storage buffer:PBS
- Storage temperature:+4 °C, –20 °C if preferred
- Concentration:1 mg/ml
- Shipping temperature:4 °C
- Purification:Purified IgG prepared by ion exchange chromatography
- Size:250 μg
- Pk:0,25 mg
Specifications
About this item
Type: Primary
Antigen: CD11B (Integrin Alpha M Chain, Mac-1)
Clonality: Monoclonal
Clone: 5C6
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse, Human