ContactUsLinkComponent
Drain stoppers for SAP and JBN baths
Drain stoppers for SAP and JBN baths
Catalog # GRANUWB-DS
Supplier:  GRANT INSTRUMENTS
undefined
Drain stoppers for SAP and JBN baths
Catalog # GRANUWB-DS
Supplier:  GRANT INSTRUMENTS
Specifications
  • For:
    JBN12, JBN18, JBN26, SAP12, SAP18, SAP26, SAP34
  • Description:
    Drain stoppers
  • Pk:
    5
Specifications
About this item
Drain stoppers, recommended when using heat transfer beads, to prevent beads entering the drain hole.