Order Entry
Belgium
Orders LinkContactUsLinkComponent
Storage Boxes, PS

Storage Boxes, PS

Clamp shell box with lid, 86×63×21 mm
Catalog # MULC41846
Supplier:   MULTIROIR
Storage Boxes, PS
Catalog # MULC41846
Supplier:  MULTIROIR
Storage Boxes, PS

Clamp shell box with lid, 86×63×21 mm

Price
242,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

PS.

  • High strength
  • Transparent flap for visibility of contents
  • With spring hinge

These boxes ensure storage of small parts safely with it's valve which closes by simply pushing.

Colour: White

Specifications
  • Description:
    Clamp shell box with lid
  • W×D×H:
    86×63×21 mm