ContactUsLinkComponent
Electrical resistant gloves, Electrosoft Composite CL 1
Electrical resistant gloves, Electrosoft Composite CL 1
Supplier:  HONEYWELL SAFETY
Certificates
undefined
Electrical resistant gloves, Electrosoft Composite CL 1
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • DALL2092017/9
  • DALL2092017/10
  • Model:
  • Composite CL1
  • Composite CL1
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex
  • Latex
  • Finish:
  • Anti-slip finish on fingers
  • Anti-slip finish on fingers
  • Cuff style:
  • Straight
  • Straight
  • EN norm:
  • EN 60903-2003
  • EN 60903-2003