Punches for sample collection cards, Whatman Uni-Core™
: QIAGEN
Manual Uni-Core™ punchers have a sharp, stainless steel cutting edge. The disposable model is designed to provide up to 500 punches cleanly and effectively. The use of the recommended punch procedure minimises the risk of sample carryover between cards. The companion cutting mat (available separately) allows for clean sample cuts and extends the life of the cutting tip.
Frequently Bought Together