Metering valves, Accuflo™
Supplier: ENTEGRIS
Certificates
About this item
These components are constructed of high purity PFA and PTFE materials for wetted parts and PVDF, PEEK™ and PTFE u-cup with Viton® energizer for non wetted parts.
- Multiple turn with manual adjustment
- Standard panel mounting configuration
- Ultrapure and corrosion-resistant performance
Maximum continuous use pressure: 689 kPa at ambient temperature.
Maximum continuous use temperature: 149 °C at 172 kPa.
Frequently Bought Together