Order Entry
Belgium
ContactUsLinkComponent
Drum pumps sets for hazardous fluids
Drum pumps sets for hazardous fluids
Catalog # LUTZ0205-411
Supplier:  LUTZ PUMPEN
undefined
Drum pumps sets for hazardous fluids
Catalog # LUTZ0205-411
Supplier:  LUTZ PUMPEN

Specifications

  • Flow rate:
    Max. 95 l/min
    Max. 60 l/min
  • Description:
    Pump set with motor ME II 3
  • Material:
    PVDF
  • Immersion depth:
    1000 mm
  • Pk:
    1 SET
  • Power:
    460 W

Specifications

About this item

This pump set is specially adapted for pumping hazardous fluids or solvents such as acetone, formic acid, butyl acetate, acetic acid, nicotine, methyl benzene (toluol) and styrol.

  • Fast assembly
  • Immediately ready for action
  • Ideal for pumping and filling thin bodied liquids
  • Compact compressed air motor powerful and reliable

The explosion proof universal motor ME II 3 is ideally suitable for transferring of many thin bodied easily flammable and combustible liquids.

Certifications: Explosion proof according to ATEX directive 94/9/EC category 2.
Delivery: Pump set includes 1 Motor ME II 3, 1 pump tube SS 41-L-SL stainless steel, 2,5 m solvent hose 1", electrically conductive bound with hose connectors brass, nozzle brass, equipotential bonding cable and drum adapter PP.