Order Entry
Belgium
ContactUsLinkComponent
 
Cell analysis slides, Arthur™
  734-2896
 :  NANOENTEK
undefined
Cell analysis slides, Arthur™
  734-2896
 :  NANOENTEK

 

  • Recommended working volume:
    25 µl
  • Description:
    Fluorescence cell counter slide
  • Material:
    PLA
  • Seq:
    901
  • Pk:
    50

 

 

Arthur™ Cell analysis slides are designed for use with the Arthur™.

  • Assay for cells including GFP&RFP expression, apoptosis, cell viability, cell cycle, and cell counting
  • Eliminates washing steps and cross contamination between samples (fully disposable)

Size: 110×24×1,9 mm