Order Entry
Belgium
Orders LinkContactUsLinkComponent
Absorptiometer cell

Absorptiometer cell

Absorptiometer cells, Glass, 28×40 mm
Catalog # TINT605534
Supplier:   Lovibond Tintometer
Absorptiometer cell
Catalog # TINT605534
Absorptiometer cell
Absorptiometer cell

Absorptiometer cells, Glass, 28×40 mm

Price
738,00 €
Log in for availability
1 * 10 ST
Product Details & Documents

Absorptiometer cell made of optical glass with path length of 10 mm.

Specifications
  • Ø×H:
    28×40 mm
  • Description:
    Absorptiometer cells
  • Material:
    Glass