Order Entry
Belgium
ContactUsLinkComponent
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 2B7]
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 2B7]
Catalog # ABNOMAB10302
Supplier:  Abnova
Anti-PRKAA1 Mouse Monoclonal Antibody [clone: 2B7]
Catalog # ABNOMAB10302
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-activated Catalytic Subunit Alpha 1
  • Antigen symbol:
    PRKAA1
  • Clonality:
    Monoclonal
  • Clone:
    2B7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    5562
  • Antigen synonyms:
    catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
  • Storage buffer:
    0,03% sodium azide
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human PRKAA1.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against recombinant PRKAA1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: PRKAA1
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat