Anti-RICK Rabbit Polyclonal Antibody
Catalog # CAYM160785-500
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:RICK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Amino acid number:11 to 30
- Sequence:PTIPYHKLADLRYLSRGASG
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human RICK amino acids 11-30
- Purification:purified by affinity chromatography
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: RICK
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: