Anti-ENHO Mouse Monoclonal Antibody [clone: CC1133F5]
Catalog # CAYM14117-200
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:ENHO
- Clonality:Monoclonal
- Clone:CC1133F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:34 to 76
- Storage buffer:PBS, pH 7,2, containing 50% glycerol and 0,02% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:human adropin amino acids 34-76
- Purification:purified by Protein G chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: ENHO
Clonality: Monoclonal
Clone: CC1133F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human