Anti-HMGB1 Mouse Monoclonal Antibody [clone: IMG19N15F4]
Catalog # CAYM11513-1
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:HMGB1
- Clonality:Monoclonal
- Clone:IMG19N15F4
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Storage buffer:PBS containing 0,05% BSA and 0,05% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:full length recombinant human HMGB1
- Purification:purified by Protein G chromatography
- Size:1 Ea
- Pk:1
Specifications
About this item
Type: Primary
Antigen: HMGB1
Clonality: Monoclonal
Clone: IMG19N15F4
Conjugation: unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse