Anti-HSL Rabbit Polyclonal Antibody
Catalog # CAYM10006371-500
Supplier: Cayman Chemical
Specifications
- Antibody type:Primary
- Antigen symbol:HSL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Amino acid number:731 to 741
- Sequence:LCRETRQAAEL
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:synthetic peptide from human HSL amino acids 731-741
- Purification:purified by affinity chromatography
- Size:500 µl
- Pk:500 µl
Specifications
About this item
Type: Primary
Antigen: HSL
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat