Order Entry
Belgium
ContactUsLinkComponent
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Catalog # VARICP3418
Supplier:  VARIAN
CAS Number:  
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Catalog # VARICP3418
Supplier:  VARIAN
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1
  • Seq:
    2370
  • Environmentally Preferable:
  • Pk Info:
    50 ml

Specifications

About this item

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol
 :
UN:3082
ADR:9,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:9002-93-1
 :9036-19-5
MDL Number:MFCD00132505