Orders LinkContactUsLinkComponent
Sample vials, with cap
   Sustainable Options Available
Supplier:   EP SCIENTIFIC
Sample vials, with cap
Supplier:  EP SCIENTIFIC
   Sustainable Options Available
Certificates
Sample vials, with cap
13 Options Available Below
Product Details & Documents
Clear or amber sample vials made from borosilicate glass.
  • Designed for water sample collection for volatile organic analysis (VOA)
  • Optimised for use with automation
  • With open-top white PP cap bonded with 3,17 mm thin PTFE-lined silicone septum
  • Septum permanently bonded to the cap for secure sealing
  • Vial shoulder slope optimized to prevent air bubble entrapment
Processed or certified to meet or exceed EPA Performance Based Specifications for volatile organic analysis with certificate. Processed vials are processed in the same manner as certified vials and come with all the features of certified vials, but vials are not barcoded and CofA is not included.
Specifications for Product Family
Specifications
  • Catalog No:
  • EPSC140-60C
  • EPSC339-20C
  • EPSC340-40C/DB
  • EPSC340-40C
  • EPSC340-60C
  • EPSCC346-0020
  • EPSCS136-0060
  • EPSCS236-0060
  • EPSCS336-0060
  • EPSCS346-0040
  • EPSC141-40A
  • EPSCS346-0060
  • EPSC339-20A
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Capacity
    Ø×H
    Colour
    Cap included
    Cap Size
    Septum
    Thickness
    Availability
    Price
  • EPSC140-60C
    Vial, VOA certified, with septa
    Vial, VOA certified, with septa
    60,00 ml
    28×139 mm
    Clear
    Yes
    24-414
    PTFE/silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 767,00 €
    767,00 €
    1 * 144 ST
       767,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC140-60C
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSC339-20C
    Vial, unprocessed, with septa
    Vial, unprocessed, with septa
    20,00 ml
    28×57 mm
    Clear
    Yes
    24-414
    PTFE/silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 166,00 €
    166,00 €
    1 * 72 ST
       166,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC339-20C
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSC340-40C/DB
    Vial, unprocessed, with septa
    Vial, unprocessed, with septa
    40,00 ml
    28×95 mm
    Clear
    Yes
    24-414
    PTFE/silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 360,00 €
    360,00 €
    1 * 144 ST
       360,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC340-40C/DB
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSC340-40C
    Vial, unprocessed, with septa
    Vial, unprocessed, with septa
    40,00 ml
    28×95 mm
    Clear
    Yes
    24-414
    PTFE/silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 204,00 €
    204,00 €
    1 * 72 ST
       204,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC340-40C
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSC340-60C
    Vial, unprocessed, with septa
    Vial, unprocessed, with septa
    60,00 ml
    28×139 mm
    Clear
    Yes
    24-414
    PTFE/silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 515,00 €
    515,00 €
    1 * 144 ST
       515,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC340-60C
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCC346-0020
    Vial, VOA certified, with closed cap
    Vial, VOA certified, with closed cap
    20,00 ml
    28×57 mm
    Amber
    Yes
    24-414
    PTFE-faced silicone, bonded
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 362,00 €
    362,00 €
    1 * 72 ST
       362,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCC346-0020
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCS136-0060
    Vial, VOA certified, with septa
    Vial, VOA certified, with septa
    60,00 ml
    28×139 mm
    Clear
    Yes
    24-414
    PTFE lined silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 280,00 €
    280,00 €
    1 * 72 ST
       280,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCS136-0060
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCS236-0060
    Vial, VOA certified, with septa
    Vial, VOA certified, with septa
    60,00 ml
    28×139 mm
    Clear
    Yes
    24-414
    PTFE lined silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 380,00 €
    380,00 €
    1 * 72 ST
       380,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCS236-0060
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCS336-0060
    Vial, VOA certified, with septa
    Vial, VOA certified, with septa
    60,00 ml
    28×139 mm
    Clear
    Yes
    24-414
    PTFE lined silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 438,00 €
    438,00 €
    1 * 72 ST
       438,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCS336-0060
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCS346-0040
    Vial, VOA certified, with septa
    Vial, VOA certified, with septa
    40,00 ml
    28×95 mm
    Amber
    Yes
    24-414
    PTFE lined silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 288,00 €
    288,00 €
    1 * 72 ST
       288,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCS346-0040
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSC141-40A
    Vial, VOA certified, with septa and PE dust cover
    Vial, VOA certified, with septa and PE dust cover
    40,00 ml
    28×95 mm
    Amber
    Yes
    24-414
    PTFE/silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 389,00 €
    389,00 €
    1 * 72 ST
       389,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC141-40A
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSCS346-0060
    Vial, VOA certified, with septa
    Vial, VOA certified, with septa
    60,00 ml
    28×139 mm
    Amber
    Yes
    24-414
    PTFE lined silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 384,00 €
    384,00 €
    1 * 72 ST
       384,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSCS346-0060
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSC339-20A
    Vial, unprocessed, with septa
    Vial, unprocessed, with septa
    20,00 ml
    28×57 mm
    Amber
    Yes
    24-414
    PTFE/silicone
    0,125"
    Log in for availability
    Log in for availability
    1 * 72 ST
    1 * 72 ST - 166,00 €
    166,00 €
    1 * 72 ST
       166,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC339-20A
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact