Anti-PR-2 Rabbit Polyclonal Antibody
Catalog # AGRIAS122366
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:PR-2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Oryza sativa
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- UniProtKB:P33157
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana PR-2 UniProt P33157, TAIR AT3G57260
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PR-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Oryza sativa