Order Entry
Belgium
ContactUsLinkComponent
 
Anti-LRRTM1 Mouse Monoclonal Antibody [clone: 2C6]
Catalog # ORIGTA805486
Supplier:  OriGene
Anti-LRRTM1 Mouse Monoclonal Antibody [clone: 2C6]
Catalog # ORIGTA805486
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    LRRTM1
  • Clonality:
    Monoclonal
  • Clone:
    2C6
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    182 to 422
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molecular weight:
    58.5 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Human recombinant protein fragment corresponding to amino acids 182-422 of human LRRTM1 (NP_849161) produced in E.coli.
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LRRTM1
Clonality: Monoclonal
Clone: 2C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human