Order Entry
Belgium
ContactUsLinkComponent
 
Toluene, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85393.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toluene, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85393.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Evaporation residue:
    Max 0.0003%
  • Colouration:
    Max 10 APHA
  • Water:
    Max 0.05%
  • GC/ECD Dioxins, Furans & PCB's:
    Max 5 ng/l
  • GC/ECD any pesticide (as Lindane):
    Max 5 ng/l
  • GC/NPD any pesticide (as Parathion):
    Max 10 ng/l
  • GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):
    Max 5 pg/ml
  • GC/ECD DCM to 1,2,4-TCB (as TCM):
    Max 1 ng/l
  • GC/FID C10 to C40 (as n-Decane):
    Max 2 ng/l
  • GC/MSD C10 to C40 (as Decane; 30-600amu):
    Max 2 ng/l
  • Filtered through 0.2um, filled under inert gas:
    Conforms

Specification Test Results

About this item

CAS Number:108-88-3
 :
Formula:C₆H₅CH₃
MDL Number:MFCD00008512
 :92.14 g/mol
Merck Index:13,09607
Boiling Pt:110.6 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.867 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under inert gas.
Flash Pt:4 °C
REACH:01-2119471310-51
 :GHS02
 :GHS08
 :GHS07