Order Entry
Belgium
ContactUsLinkComponent
2-Propanol ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Flammable
Irritant
2-Propanol ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85391.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
2-Propanol ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85391.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.9 %
  • Evaporation residue:
    Max 0.0003%
  • Colouration:
    Max 10 APHA
  • Water:
    Max 0.05%
  • GC/ECD Dioxins, Furans & PCB's:
    Max 5 ng/l
  • GC/ECD any pesticide (as Lindane):
    Max 5 ng/l
  • GC/NPD any pesticide (as Parathion):
    Max 10 ng/l
  • GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):
    Max 5 pg/ml
  • GC/ECD DCM to 1,2,4-TCB (as TCM):
    Max 2 ng/l
  • GC/FID C10 to C40 (as n-Decane):
    Max 2 ng/l
  • GC/MSD C10 to C40 (as Decane; 30-600amu):
    Max 2 ng/l
  • Filtered through 0.2um, filled under inert gas:
    Conforms

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under inert gas.
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674