Order Entry
Belgium
ContactUsLinkComponent
n-Hexane ≥99%, PESTINORM® Supra Trace for organic trace analysis
n-Hexane ≥99%, PESTINORM® Supra Trace for organic trace analysis
Supplier:  VWR Chemicals
Certificates
undefined
n-Hexane ≥99%, PESTINORM® Supra Trace for organic trace analysis
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99 %
  • Evaporation residue:
    Max 0.0003%
  • Colouration:
    Max 10 APHA
  • Water:
    Max 0.005%
  • GC/ECD Dioxins, Furans & PCB's:
    Max 2 ng/l
  • GC/ECD any pesticide (as Lindane):
    Max 2 ng/l
  • GC/NPD any pesticide (as Parathion):
    Max 5 ng/l
  • GC/ECD 1,2,4-TCB to daca-PCB (as Lindane):
    Max 2 pg/ml
  • GC/ECD DCM to 1,2,4-TCB (as TCM):
    Max 1 ng/l
  • GC/FID C10 to C40 (as n-Decane):
    Max 2 ng/l
  • GC/MSD C10 to C40 (as Decane; 30-600amu):
    Max 2 ng/l
  • Filtered through 0.2um, filled under inert gas:
    Conforms

Specification Test Results

Related Information

Frequently Bought Together