Orders LinkContactUsLinkComponent
Screw caps
Screw caps
   Sustainable Options Available
Supplier:   EP SCIENTIFIC
Screw caps
Supplier:  EP SCIENTIFIC
   Sustainable Options Available
Certificates
Screw caps
3 Options Available Below
Product Details & Documents
LDPE-lined white caps.
  • For general-purpose HDPE bulk separate containers
  • Same high quality caps used on certified HDPE environmental sample containers
Bestelinformatie: Bottles sold separately (IR013-series and IR012-series).
Specifications for Product Family
Specifications
  • Catalog No:
  • EPSC067-2410WBP
  • EPSC067-3800WBP
  • EPSC067-4800WBP
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Thread
    Availability
    Price
  • EPSC067-2410WBP
    Caps, PP, white, LDPE foam lined
    Caps, PP, white, LDPE foam lined
    24-410
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 144,00 €
    144,00 €
    1 * 250 ST
       144,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC067-2410WBP
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSC067-3800WBP
    Caps, PP, white, LDPE foam lined
    Caps, PP, white, LDPE foam lined
    38-400
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 332,00 €
    332,00 €
    1 * 500 ST
       332,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC067-3800WBP
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • EPSC067-4800WBP
    Caps, PP, white, LDPE foam lined
    Caps, PP, white, LDPE foam lined
    48-400
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 173,00 €
    173,00 €
    1 * 250 ST
       173,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    EPSC067-4800WBP
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.