Bottle racks
Supplier: MERCK LIFE SCIENCE
Jump to:
About this item
Racks for media bottles, made of high quality stainless steel, electrolytic polished.
- Autoclavable
- Easy to clean
- Good chemical and temperature resistance
Safe and convenient transport of bottles. Suitable for isolators and clean rooms.
2 Options Available Below
Product Details & Documents
Racks for media bottles, made of high quality stainless steel, electrolytic polished.
- Autoclavable
- Easy to clean
- Good chemical and temperature resistance
Safe and convenient transport of bottles. Suitable for isolators and clean rooms.
Product Family Options
Product Information
- AvailabilityPrice
- HEIP1465260001Bottle rack, 15× 100 ml (max. 50 mm diameter)
- HEIP1465250001Bottle rack, 6× 1 litre (max. 100 mm diameter)