Multiple gradient gel caster, Owl™
Gel caster, for gel size 250×130×230 mm (W×D×H)
Catalog # 730-0296
Supplier: THERMO OWL SCIENTIFIC
Jump to:
Product Details & Documents
Multiple gradient gel caster for casting one to five gels simultaneously.
Specifications
- Description:Gel caster, for gel size 250×130×230 mm (W×D×H)
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement clamps