ContactUsLinkComponent
Bottles, coliform water sample
Bottles, coliform water sample
Catalog # EPSC156-120ST
Supplier:  EP SCIENTIFIC
   Low Manufacturing Impact
undefined
Bottles, coliform water sample
Catalog # EPSC156-120ST
Supplier:  EP SCIENTIFIC
   Low Manufacturing Impact
Specifications
  • Packed:
    Case of 200
  • Capacity:
    120 ml
  • Material:
    PS
  • Seq:
    02
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    200
  • Dosage:
    Undosed
Specifications
About this item
PP, PE or PS.
  • Certified sterile; manufactured under an ISO certified quality management system
  • Threaded screw closure for easy aseptic sample collection and secure cap engagement
  • Non-autofluorescent
  • Available pre-charged with a sodium thiosulfate (Na₂S₂O₃) tablet for sample de-chlorination, or without a tablet for un-chlorinated water sources
  • 100 ml fill line
Designed for use with EPA compliant total coliform and fecal coliform water analysis using the Colilert analysis method or equivalent.