Order Entry
Belgium
ContactUsLinkComponent
Dichloromethane ≥99.9% stabilised, PESTINORM® Supra Trace for organic trace analysis
Health
Irritant
Dichloromethane ≥99.9% stabilised, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85386.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Dichloromethane ≥99.9% stabilised, PESTINORM® Supra Trace for organic trace analysis
Catalog # 85386.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Assay (calculated on anhydrous):
    Min. 99.9 %
  • Identity (IR):
    Conforms
  • Residue after evaporation:
    Max. 0.0003 %(m)
  • Water:
    Max. 0.005 %(m)
  • Colour:
    Max. 10 APHA
  • GC/ECD dioxins, furans and PCB's:
    Max. 2.0 ng/l
  • GC/ECD any pesticide (as Lindane):
    Min. 2 ng/l
  • GC/NPD any pesticide (as Parathion):
    Max. 5 ng/l
  • GC/ECD 1,2,4-TCB to deca-PCB(as Lindane):
    Max. 2.0 pg/ml
  • GC/FID C10 to C40 (as n-Decane):
    Max. 2.0 ng/ml
  • GC/MSD C10 to C40 (as Decane; 30-600amu):
    Max. 2.0 ng/ml
  • Stabilizer (Amylene):
    0.002 - 0.006 %(m)

Specification Test Results

About this item

CAS Number:75-09-2
 :
Formula:CH₂Cl₂
MDL Number:MFCD00000881
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :Filtered through a 0.2 µm filter, packaged under inert gas.
 :Stabilised with amylene 0.002-0.006 %
REACH:01-2119480404-41
 :GHS08
 :GHS07
Storage Temperature:Ambient