Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) ≥99.8%, PESTINORM® for headspace gas chromatography
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) ≥99.8%, PESTINORM® for headspace gas chromatography
Catalog # 85398.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) ≥99.8%, PESTINORM® for headspace gas chromatography
Catalog # 85398.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Purity (GC):
    min. 99.8%
  • Identity (IR):
    Complying
  • Water (KF):
    max. 0.05%w/w
  • Color (APHA Pt-Co):
    max. 10
  • GC-HS (Each class 1 solvent):
    max. 1µg/g
  • GC-HS (Each class 2 solvent):
    max. 10µg/g
  • GC-HS (Each class 3 solvent):
    max. 50µg/g
  • GC-HS (Each unknown, MIBK-DMSO):
    max. 10µg/g
  • Suitable as solvent for the analysis of residual solvents ofICH classes 1, 2 and 3 according to Ph Eur and USP.:
    Passes test

Specification Test Results

About this item

 :
 :99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
Storage Temperature:Ambient
EINECS:212-828-1
REACH:01-2119472430-46
 :GHS08
 :GHS07
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193