Orders LinkContactUsLinkComponent
Separating funnels
Separating funnels
Supplier:   QVF LABORTECHNIK
Separating funnels
Supplier:  QVF LABORTECHNIK
Certificates
Separating funnels
3 Options Available Below
Product Details & Documents
Made of borosilicate glass.
  • Stopcock with glass key
  • Without graduations
  • Excellent chemical and temperature resistance
The separating funnels are used for discontinuous shaking out of solvents.
Certifications: Complies to DIN 12 451.
Product Family Options
Product Information
  • Capacity
    Socket size
    Availability
    Price
  • 50 ml
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • 250 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 125,00 €
    125,00 €
    1 * 1 ST
       125,00 €
  • 2000 ml
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST