Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

pH Buffer solutions

Supplier:   Hach
pH Buffer solutions
Supplier:  Hach
Certificates
pH Buffer solutions
pH Buffer solutions

7 Options Available Below

Product Details & Documents

For calibration of pH meters.

Accuracy: pH is ±0,02 at 25 °C

 : Traceable to NIST.

 : Please note that the pH 4 and 10 buffers contain formaldehyde <0,1% and the pH 7 buffer contains formaldehyde 0,1 to 1%.

Product Family Options

Product Information

  • pH value
    Colour
    Pk
    Availability
    Price
  • HACHS11M009
    Buffer solution, technical grade
    Buffer solution, technical grade
    1,09
    Colourless
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 39,70 €
    39,70 €
    1 * 500 mL
       39,70 €

  • Buffer solution
    4,01
    Colourless
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 23,00 €
    23,00 €
    1 * 500 mL
       23,00 €

  • HACHS11M010
    Buffer solution, technical grade
    Buffer solution, technical grade
    4,65
    Colourless
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 39,70 €
    39,70 €
    1 * 500 mL
       39,70 €

  • Buffer solution
    7,00
    Colourless
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 23,00 €
    23,00 €
    1 * 500 mL
       23,00 €

  • HACH1222220
    Buffer solution
    Buffer solution
    7,00
    Colourless
    25 mL
    Log in for availability
    Log in for availability
    1 * 25 mL
    1 * 25 mL - 21,50 €
    21,50 €
    1 * 25 mL
       21,50 €

  • HACHS11M011
    Buffer solution, technical grade
    Buffer solution, technical grade
    9,23
    Colourless
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 39,70 €
    39,70 €
    1 * 500 mL
       39,70 €

  • Buffer solution
    10,00
    Colourless
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 23,00 €
    23,00 €
    1 * 500 mL
       23,00 €