Anti-IgG Antibody (Biotin)
BSBTBA1020-0.5
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Antigen symbol:IgG
- Conjugation:Biotin
- Host:
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Size:500 μg
- Pk:0,5 mg
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity: