Orders LinkContactUsLinkComponent
Cylindrical bend testers
Cylindrical bend testers
Supplier:   Industrial Physics
Cylindrical bend testers
Supplier:  Industrial Physics
Certificates
Cylindrical bend testers
2 Options Available Below
Product Details & Documents
These cylindrical bend testers are used to determine the elasticity, adhesion and elongation of paint on metal.
  • Sturdy conception in anodised aluminium and stainless steel
  • Ergonomic clamping device for test panels
  • Large knob on bending arm for smooth bending
The test panel is bent over the cylindrical mandrel and then checked for damages.
Certifications: ISO 1519 (or ASTM D522)
Delivery information: Supplied with a desktop holder and mandrels.
Documents
Product Family Options
Product Information
  • Availability
    Price
  • SIMESP1820
    Cylindrical bend tester (100 mm) with desk holder and 14 mandrels (Ø = 2 - 32 mm)
    Cylindrical bend tester (100 mm) with desk holder and 14 mandrels (Ø = 2 - 32 mm)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 260,00 €
    2 260,00 €
    1 * 1 ST
       2 260,00 €
  • SIMESP1822
    Cylindrical bend tester (100 mm) with desk holder and seven mandrels (Ø = 1/8 to 1") 
    Cylindrical bend tester (100 mm) with desk holder and seven mandrels (Ø = 1/8 to 1") 
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 310,00 €
    2 310,00 €
    1 * 1 ST
       2 310,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 6 mm
    Mandrel, 6 mm
    SIMESP1840
    Log in for availability
    1 * 1 ST - 69,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 10 mm
    Mandrel, 10 mm
    SIMESP1842
    Log in for availability
    1 * 1 ST - 69,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Set of 14 mandrels (Ø = 2, 3, 4, 5, 6, 8, 10, 12, 13, 16, 19, 20, 25, 32 mm) with desktop holder
    Set of 14 mandrels (Ø = 2, 3, 4, 5, 6, 8, 10, 12, 13, 16, 19, 20, 25, 32 mm) with desktop holder
    SIMESP1835
    Log in for availability
    1 * 14 ST - 543,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 2 mm
    Mandrel, 2 mm
    SIMESP1836
    Log in for availability
    1 * 1 ST - 69,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 3 mm
    Mandrel, 3 mm
    SIMESP1837
    Log in for availability
    1 * 1 ST - 69,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 4 mm
    Mandrel, 4 mm
    SIMESP1838
    Log in for availability
    1 * 1 ST - 69,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 5 mm
    Mandrel, 5 mm
    SIMESP1839
    Log in for availability
    1 * 1 ST - 59,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 12 mm
    Mandrel, 12 mm
    SIMESP1843
    Log in for availability
    1 * 1 ST - 69,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 8 mm
    Mandrel, 8 mm
    SIMESP1841
    Log in for availability
    1 * 1 ST - 59,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 13 mm
    Mandrel, 13 mm
    SIMESP1844
    Log in for availability
    1 * 1 ST - 59,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 16 mm
    Mandrel, 16 mm
    SIMESP1845
    Log in for availability
    1 * 1 ST - 69,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 19 mm
    Mandrel, 19 mm
    SIMESP1846
    Log in for availability
    1 * 1 ST - 59,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 20 mm
    Mandrel, 20 mm
    SIMESP1847
    Log in for availability
    1 * 1 ST - 69,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 25 mm
    Mandrel, 25 mm
    SIMESP1848
    Log in for availability
    1 * 1 ST - 59,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 32 mm
    Mandrel, 32 mm
    SIMESP1849
    Log in for availability
    1 * 1 ST - 59,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Set of seven mandrels (Ø = 1/8, 1/4, 3/8, 1/2, 5/8, 3/4, 1") with desktop holder
    Set of seven mandrels (Ø = 1/8, 1/4, 3/8, 1/2, 5/8, 3/4, 1") with desktop holder
    SIMESP1833
    Log in for availability
    1 * 1 ST - 549,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 3/4"
    Mandrel, 3/4"
    SIMESP1828
    Log in for availability
    1 * 1 ST - 196,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 1/8"
    Mandrel, 1/8"
    SIMESP1823
    Log in for availability
    1 * 1 ST - 153,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 1/4"
    Mandrel, 1/4"
    SIMESP1824
    Log in for availability
    1 * 1 ST - 196,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 3/8"
    Mandrel, 3/8"
    SIMESP1825
    Log in for availability
    1 * 1 ST - 191,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 1/2"
    Mandrel, 1/2"
    SIMESP1826
    Log in for availability
    1 * 1 ST - 153,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Mandrel, 5/8"
    Mandrel, 5/8"
    SIMESP1827
    Log in for availability
    1 * 1 ST - 191,00 €