ContactUsLinkComponent
Aspirating pipettes
Aspirating pipettes
Catalog # GREI710183_1000
Supplier:  Greiner Bio-One
undefined
Aspirating pipettes
Catalog # GREI710183_1000
Supplier:  Greiner Bio-One
Specifications
  • Capacity:
    2 ml
  • Pk:
    1.000
Specifications
About this item
PS pipettes, non plugged, non graduated, disposable, sterile.
  • Free of detectable DNase, RNase, human DNA
  • Non pyrogenic, non cytotoxic
  • Maximum accuracy, drop-free pipetting
  • Maximum clarity from high-grade polystyrene
  • Lot number and best-before date on each bag
Packaging: Paper/plastic-packaging with peel-off function for individually wrapped pipettes with an additional break-through function.