ContactUsLinkComponent
Wipes for general applications, Polytex® light
Wipes for general applications, Polytex® light
  ZVGA10160-00
undefined
Wipes for general applications, Polytex® light
  ZVGA10160-00
  • Packed:
    Pack of 420 (12×35 wipes)
  • Colour:
    White
  • L×W:
    400×420 mm
  • Pk:
    420
These solvent resistant wipes made of 100% PP are suitable for wet wiping in a variety of applications.
  • Durable and lint-free
  • Even remove stubborn dirt
  • Suitable for food contact
Ideal for use in garages, paint shops, catering areas, kitchens and food processing areas.