Capillary columns, DB-EUPAH
Supplier: Agilent
Aller à :
À propos de cet article
Polycyclic aromatic hydrocarbons are chemical compounds consisting of fused aromatic rings formed during the incomplete combustion of organic materials.
- With excellent signal-to-noise ratio
- Optimised column dimensions for better performance
- Great resolution of critical isomers
- Good thermal stability
2 Options disponibles ci-dessous
Product Details & Documents
Polycyclic aromatic hydrocarbons are chemical compounds consisting of fused aromatic rings formed during the incomplete combustion of organic materials.
- With excellent signal-to-noise ratio
- Optimised column dimensions for better performance
- Great resolution of critical isomers
- Good thermal stability
Product Family Options
Informations sur le produit
- Column FormatPhaseFilm thicknessØ int.LengthAvailabilityPrice
- HEWL121-9627on 7" cage
- HEWL122-96L2on 7" cage