ContactUsLinkComponent
behrotest® Clean-up station, CUS 2
behrotest® Clean-up station, CUS 2
Catalog # BEHRB00217638
Supplier:  BEHR
undefined
behrotest® Clean-up station, CUS 2
Catalog # BEHRB00217638
Supplier:  BEHR
Specifications
  • Description:
    Clean-up station
  • Pk:
    1
  • Type:
    CUS 2
Specifications
About this item
The complete sample clean up station includes dropping funnel, clean-up column with glass frit, reducing flasks and stand.
The clean up procedure can be done for two samples simultaneously.
Certifications: Corresponds exactly to the provisions of ISO 9377-2.