ContactUsLinkComponent
Racks for SPB system tubes
Racks for SPB system tubes
Catalog # PERCN9308070
Supplier:  PerkinElmer
Racks for SPB system tubes
Catalog # PERCN9308070
Supplier:  PerkinElmer
Specifications
  • For tubes:
    50 ml
  • No. of Holes:
    24 (4×6)
  • Description:
    Rack for SPB 50-24 system
  • Pk:
    1
Specifications
About this item
Polycarbonate.
  • With rack lock feature which locks DigiTUBE®s in position within the rack allowing for easy and one-handed closure of screw cap
  • Acid resistant
  • For laboratory usage