Plane weighing glass, according to Heidbrink
611-9090
- Description:Plane weighing glass acc. to Heidbrink
- Environmentally Preferable:
- Pk:1
Borosilicate glass.
- Excellent chemical resistance
- Excellent temperature resistance
For all weighings in which there is an advantage when the sample is divided in thin layers.