EnteroPluri test
About this item
EnteroPluri Test is a 12-sector system containing special culture media that permits identification of the Enterobacteriaceae and other gram negative, oxidase negative bacteria. The system allows the simultaneous inoculation of all media present in the sectors and the execution of 15 biochemical reactions. Microorganism is identified evaluating the colour change of the different culture media after 18 to 24 hours of incubation at 36 ±1 °C and by a code number obtained from biochemical reaction interpretation. Oxidase, Kovacs Reagent Indol (e.g. 1.00181.0002, 1.09293.0100, 1.11350.0001) and Codebook are not included.
- Based on biochemical tests performed on culture media containing specific substrates
- Combination of positive and negative permits identification of bacteria by using the Codebook
17 Options Available Below
Product Details & Documents
EnteroPluri Test is a 12-sector system containing special culture media that permits identification of the Enterobacteriaceae and other gram negative, oxidase negative bacteria. The system allows the simultaneous inoculation of all media present in the sectors and the execution of 15 biochemical reactions. Microorganism is identified evaluating the colour change of the different culture media after 18 to 24 hours of incubation at 36 ±1 °C and by a code number obtained from biochemical reaction interpretation. Oxidase, Kovacs Reagent Indol (e.g. 1.00181.0002, 1.09293.0100, 1.11350.0001) and Codebook are not included.
- Based on biochemical tests performed on culture media containing specific substrates
- Combination of positive and negative permits identification of bacteria by using the Codebook
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- PkAvailabilityPrice
- LIOF71618Enterosystem 18 R
- LIOF71640Listeria system 18R
- LIOF71708Bacterial Code Book for Enterobacteriaceae identification using Oxi/Ferm Pluri TestBacterial Code Book for Enterobacteriaceae identification using Oxi/Ferm Pluri Test1Log in for availability1 * 1 ST39,80 €
- LIOF71709EnteroPluri-Test Codebook, bacterial code book for Enterobacteriaceae identificationEnteroPluri-Test Codebook, bacterial code book for Enterobacteriaceae identification1Log in for availability1 * 1 ST41,20 €
- LIOF72560Strepto system 12R
- LIOF78618EnteroPluri-Test, system with 12 agar media by simultaneous inoculum for Enterobacteriaceae identificationEnteroPluri-Test, system with 12 agar media by simultaneous inoculum for Enterobacteriaceae identification10 TestsLog in for availability1 * 1 EA105,00 €
- LIOF78619EnteroPluri-Test, system with 12 agar media by simultaneous inoculum for Enterobacteriaceae identificationEnteroPluri-Test, system with 12 agar media by simultaneous inoculum for Enterobacteriaceae identification25 TestsLog in for availability1 * 25 Tests240,00 €
- LIOF78620System for the identification of Gram negative, oxidase positive bacteriaSystem for the identification of Gram negative, oxidase positive bacteria1Log in for availability1 * 1 ST105,00 €
- LIOF78621System for the identification of Gram negative, oxidase positive bacteriaSystem for the identification of Gram negative, oxidase positive bacteria1Log in for availability1 * 1 ST240,00 €
- LIOF80252Additional biochemical reagents for Entero System 18 RAdditional biochemical reagents for Entero System 18 R1 KITLog in for availability1 * 1 KIT76,90 €
- LIOF80257Additional biochemical reagents for Listeria System 18Additional biochemical reagents for Listeria System 181 KITLog in for availability1 * 1 KIT113,00 €
- LIOF80260Additional biochemical reagents for Staph identification systemsAdditional biochemical reagents for Staph identification systems1 KITLog in for availability1 * 1 KIT141,00 €
- LIOF87002V.P. TEST Reagent and NaOH and alpha-naphthol solutions for Voges Proskauer testV.P. TEST Reagent and NaOH and alpha-naphthol solutions for Voges Proskauer test1 KITLog in for availability1 * 1 KIT102,00 €
- LIOF71710“Code book” manual for Enterosystem 18R
- LIOF71619Enterosystem 24R: Biochemical identification of enterobacteriaEnterosystem 24R: Biochemical identification of enterobacteriaLog in for availability1 * 1 ST173,00 €
- LIOF71620Anaerobe system - biochemical identification of anaerobic microorganismsAnaerobe system - biochemical identification of anaerobic microorganismsLog in for availability1 * 20 ST225,00 €
- LIOF79618Enterosystem 18R: Biochemical identification of enterobacteriaEnterosystem 18R: Biochemical identification of enterobacteriaLog in for availability1 * 4 Tests32,90 €
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.