Orders LinkContactUsLinkComponent
Latex tests
Latex tests
Test serum for Yersina enterocolitica identification, 1
N° de catalogue LIOF96147
Fournisseur:   LIOFILCHEM
Latex tests
N° de catalogue LIOF96147
Fournisseur:  LIOFILCHEM
Numéro CAS:  
Latex tests
Latex tests
Test serum for Yersina enterocolitica identification, 1
Numéro CAS:  
Prix
260,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Identification of the main pathogenic microorganisms directly from clinical specimens.
  • Save time and labor with direct testing
  • Results in 18 to 24 hours
  • Confirmation with latex agglutination tests
  • Clear and well defined colour changes
  • Easy interpretation
Documents
Spécifications
Spécifications
  • Pk:
    1
  • Description:
    Test serum for Yersina enterocolitica identification
Spécifications