ContactUsLinkComponent
Beakers, with handle, porcelain
Beakers, with handle, porcelain
Catalog # HECH47020030
Supplier #: 47020030
undefined
Beakers, with handle, porcelain
Catalog # HECH47020030
Supplier #: 47020030
Log in for availability
Select an Option
100 ml
Select an Option
02
Select an Option
1
About this item
Porcelain, glazed white.
  • Exceptional chemical resistance
  • Excellent thermal resistance
  • Robust, heavy construction
Cylindrical, with spout. With very easy-to-read, internal graduations.
Frequently Bought Together