Order Entry
Belgium
Orders LinkContactUsLinkComponent
Base materials for microbiology media

Base materials for microbiology media

Base materials for microbiology media
Supplier:  EUROCLONE
Certificates
Base materials for microbiology media
Base materials for microbiology media

11 Options Available Below

Product Details & Documents

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • EURLEMR250001
    Yeast extract
    Yeast extract
    Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

  • EURLEMR250005
    Yeast extract
    Yeast extract
    Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

  • EURLEMR250100
    Yeast extract
    Yeast extract
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • EURLEMR250250
    Yeast extract
    Yeast extract
    Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g

  • EURLEMR250500
    Yeast extract
    Yeast extract
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • MOLE63514852-500G
    Casein hydrolysate, enzymatic digest
    Casein hydrolysate, enzymatic digest
    Plastic bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
  • MOLE35752308-1KG
    Yeast extract
    Yeast extract
    Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

    Specifications:
    Marketsource
    Marketsource
  • Tryptone
    Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
  • IMPA8534072
    Agar No. 1 bacteriological high clarification
    Agar No. 1 bacteriological high clarification
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
  • IMPA8534161
    Bacteriological peptone
    Bacteriological peptone
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g

    Specifications:
    Marketsource
    Marketsource
  • Proteose peptone
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 119,00 €
    119,00 €
    1 * 500 g
       119,00 €

This product has been blocked by your organization. Please contact your purchasing department for more information
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.